Learn More
System Biosciences LLC EXOSOME-DEPLTED FBS Exosome-depleted FBS Media Supplement = 250ml
SDP
Catalog No. NC0464480
Encompass_Preferred
Bovine exosome-depleted FBS, for isolating exosomes from cultured cells without the complication of bovine exosomes that are present in conventional FBS. Fetal bovine serum, or FBS, is an important component of many types of cell culture media. But for researchers interested in isolating exosomes from cultured cells, standard FBS can introduce unwanted complications—bovine exosomes, which can cause significant background issues or interfere with functional studies. Which is why SBI developed Exo-FBS, our patented exosome-depleted FBS.
Catalog No. NC0464480 Supplier System Biosciences LLC Supplier No. EXOFBS250A1
Bovine exosome-depleted FBS, for isolating exosomes from cultured cells without the complication of bovine exosomes that are present in conventional FBS. Fetal bovine serum, or FBS, is an important component of many types of cell culture media. But for researchers interested in isolating exosomes from cultured cells, standard FBS can introduce unwanted complications—bovine exosomes, which can cause significant background issues or interfere with functional studies. Which is why SBI developed Exo-FBS, our patented exosome-depleted FBS.