Learn More
Neuromics, Inc Exosome Free Fetal Bovine Serum, 50 ml
Catalog No. NC2071929
Encompass
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance.Our Exosome Depleted FBS features: High-level Exosome depletion, Endotoxin levels : ≤10 EU/mL, Triple 0.1 μm Filtered, Free of Virus and Mycoplasma.Exosome Depleted FBS removes Exosomes while retaining growth nutrients for cell culturing. Excellent for research applications where Exosomes cause complications.
Catalog No. NC2071929 Supplier Neuromics, Inc Supplier No. FBS002X50
Our FBS is manufactured in a fully integrated environment from raw material collection to finished product. It is produced to the highest standards in order to reduce risk and product variability, and to ensure product safety and performance.Our Exosome Depleted FBS features: High-level Exosome depletion, Endotoxin levels : ≤10 EU/mL, Triple 0.1 μm Filtered, Free of Virus and Mycoplasma.Exosome Depleted FBS removes Exosomes while retaining growth nutrients for cell culturing. Excellent for research applications where Exosomes cause complications.