Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Exosome Standard (HCT116 cell line)

Catalog No. 89998623 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 x 30 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-623 2 x 30 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-623 Supplier Abnova™ Supplier No. U0350

This product is purified and lyophilized exosomes obtained from HCT116 cell line (human colon carcinoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.

  • Reconstitute lyophilized exosome standard by adding deionized water. 30 μL or 100 μL for lyophilized standard to get a final concentration of 1 μg/μL. Different volumes of deionized water for exosomes reconstitution can be chosen by the users in according with the desired final concentration. Suspend exosomes pipetting the solution up and down 10-15 times and avoid bubbles. Vortex the reconstituted standard for 60 seconds. Briefly centrifuge the tubes containing the standard to ensure that the solution is collected at the bottom of the tube. Pipette the solution up and down 10 times, avoiding the introduction of bubbles. After this step, the standard is ready to use.
  • Exosomes are purified following a combination of ultracentrifugation and microfiltration steps. Exosomes are subsequently quantified and validated for overall protein content and particle number by NTA (Nanoparticles Tracking Analysis). Lyophilization does not affect the stability of purified exosomes and the expression levels of their exosome markers (proteins and nucleic acids).
  • Lyophilized exosome standards can be stored at 4°C for up to 36 months. Reconstituted exosome standards are not suitable for long-term storage at room temperature, use them within 2 hours after reconstitution. The remaining reconstituted standard stock solution should be aliquoted into low binding polypropylene vials and stored at -20°C for up to 1 month or at -80°C for up to 6 months. Strictly avoid repeated freeze-and-thaw cycles.
  • Raising in host species: Human
  • Quality control testing: Comparison of exosomal markers on fresh and lyophilized exosomes
  • Type clonality: Protein
  • Applications: Western Blotting, Elisa, Flow Cytometry

Specifications

Content And Storage Store at 4°C for up to 36 months
Description Storage Buffer: Lyophilized from cell culture supernatant
Lyophilized Yes
Quantity 2 x 30 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.