Learn More
System Biosciences LLC HEK293 EXOSOMESFrozen exosomes (>1x10^10) from HEK293 Human embryonic kidney cell line = 50ug
SDP
Catalog No. NC1785507
Encompass_Preferred
Save time with our ready-to-use purified exosomes Whether you’re looking for exosome standards, performing functional studies, evaluating biomarkers, or engineering exosomes for therapeutic delivery, you can get your research moving faster with our ready-to-use purified exosomes. Prepared with quality and care Like all of our purified exosomes, exosomes isolated from HEK293 cells are carefully manufactured in SBI’s Palo Alto facilities. Starting with cells grown in Exo-FBS (our exosome-depleted FBS) to maximize purity, exosomes are isolated using ExoQuick-TC and then quality checked by Western blot for the presence of CD63, and NanoSight for particle size and intactness. Provided in amounts > 1 x 106 exosomes (or 50 µg protein equivalent), all of our preps deliver functional exosomes.
Catalog No. NC1785507 Supplier System Biosciences LLC Supplier No. EXOP110A1
Save time with our ready-to-use purified exosomes Whether you’re looking for exosome standards, performing functional studies, evaluating biomarkers, or engineering exosomes for therapeutic delivery, you can get your research moving faster with our ready-to-use purified exosomes. Prepared with quality and care Like all of our purified exosomes, exosomes isolated from HEK293 cells are carefully manufactured in SBI’s Palo Alto facilities. Starting with cells grown in Exo-FBS (our exosome-depleted FBS) to maximize purity, exosomes are isolated using ExoQuick-TC and then quality checked by Western blot for the presence of CD63, and NanoSight for particle size and intactness. Provided in amounts > 1 x 106 exosomes (or 50 µg protein equivalent), all of our preps deliver functional exosomes.