Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Exothermic Reactats
SCI_ED

Catalog No. s96736 Shop All Aldon Products
Change view
Click to view available options
Exothermic Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S96736 Exothermic Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S96736 Supplier Innovating Science™ Supplier No. IS7004
This item is not returnable. View return policy

Includes

Materials for five demonstrations: Bags (5); 25 g Iron Metal powder, UN 3089; 1 g Sodium Chloride crystals; 5 g Calcium Chloride flakes; 1 Tbsp. Vermiculite; and instructions

Study the process of exothermic reactions.

  • Reaction is safely carried out in a plastic bag
  • Students can pass bag around to feel reaction

Compliance

Package conforms to 49 CFR 173.4

Specifications

Certifications/Compliance 49 CFR 173.4
Product Type Exothermic Reactions Demonstration Kit
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Study the process of exothermic reactions
Includes 5 bags of the following mixture 25 g iron metal powder, 1 g sodium chloride, crystals, 5 g calcium chloride flakes, 1 tbsp vermiculite
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 5 Demonstrations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.