Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Exothermic Reactions
GREENER_CHOICESCI_ED

Catalog No. S96894 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S96894 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96894 Supplier Innovating Science™ Supplier No. IS2501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Materials for 15 groups: 100g Calcium Chloride; plastic cups (15); stirring sticks (15); 15 bags with: 25gIron powder UN3089,; 1g Sodium Chloride, 5g Calcium Chloride,and 1 Tbsp. Vermiculite™

See how chemistry can be used in a beneficial manner

Investigate temperature change of the reaction and examine how the oxidation of iron can be used to create heat packs. Enough materials for 15 groups.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To investigate temperature change of the reaction and examine how the oxidation of iron can be used to create heat packs
Includes 1 x 100g calcium chloride, 15 plastic cups, 15 stirring sticks, 15 bags containing 25g iron powder, 1g sodium chloride, 5g calcium chloride, 1 table spoon vermiculite, teacher's manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Exothermic Reaction Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.