Learn More
Eurofins Discoverx PathHunter eXpress ACVRL1/ACVR2B Dimerization Assay, 10pl Kit. (93-0964E3CP22L)
Catalog No. 502001689
Encompass
The PathHunter ACVRL1/ACVR2B Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. GDF2/BMP9) induced dimerization of ACVRL1/ACVR2B receptors. This product uses EFC technology. The cell line included overexpresses ACVRL1 target protein fused to PK alongside ACVR2B target protein fused to EA. Upon ligand-induced dimerization of ACVRL1/ACVR2B, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1689 Supplier Eurofins Discoverx Supplier No. 930964E3CP22L
Add to Cart
Add to Cart
Request a Quote
The PathHunter ACVRL1/ACVR2B Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. GDF2/BMP9) induced dimerization of ACVRL1/ACVR2B receptors. This product uses EFC technology. The cell line included overexpresses ACVRL1 target protein fused to PK alongside ACVR2B target protein fused to EA. Upon ligand-induced dimerization of ACVRL1/ACVR2B, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.