Learn More
Eurofins Discoverx cAMP Hunter eXpress ADORA2B CHO-K1 GPCR Assay, 2-Plate Kit. (95-0041E2CP2M)
Catalog No. 502002296
Encompass
The cAMP Hunter ADORA2B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. NECA) based activation of ADORA2B GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type ADORA2B, a naturally Gαs-coupled GPCR. Upon activation of ADORA2B, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2296 Supplier Eurofins Discoverx Supplier No. 950041E2CP2M
The cAMP Hunter ADORA2B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. NECA) based activation of ADORA2B GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type ADORA2B, a naturally Gαs-coupled GPCR. Upon activation of ADORA2B, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.