Learn More
Eurofins Discoverx PathHunter eXpress ADRA1B CHO-K1 β-Arrestin GPCR Assay, 2-Plate Kit. (93-0274E2CP0M)
Catalog No. 502000148
Encompass
The PathHunter ADRA1B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA1B and EA-tagged β-Arrestin-2. Activation of ADRA1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0148 Supplier Eurofins Discoverx Supplier No. 930274E2CP0M
The PathHunter ADRA1B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA1B and EA-tagged β-Arrestin-2. Activation of ADRA1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.