Learn More
Eurofins Discoverx PathHunter eXpress ADRA2B CHO-K1 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0641E2CP2L)
Catalog No. 502001014
Encompass
The PathHunter ADRA2B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA2B and EA-tagged β-Arrestin-2. Activation of ADRA2B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1014 Supplier Eurofins Discoverx Supplier No. 930641E2CP2L
Add to Cart
Add to Cart
Request a Quote
The PathHunter ADRA2B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA2B and EA-tagged β-Arrestin-2. Activation of ADRA2B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.