Learn More
Eurofins Discoverx cAMP Hunter eXpress ADRA2C CHO-K1 GPCR Assay, 2-Plate Kit. (95-0053E2CP2M)
Catalog No. 502002505
Encompass
The cAMP Hunter ADRA2C eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2C GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type ADRA2C, a naturally Gi/Go coupled GPCR. Upon activation of ADRA2C, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2505 Supplier Eurofins Discoverx Supplier No. 950053E2CP2M
The cAMP Hunter ADRA2C eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. epinephrine and norepinephrine) based activation of ADRA2C GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type ADRA2C, a naturally Gi/Go coupled GPCR. Upon activation of ADRA2C, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.