Learn More
Eurofins Discoverx PathHunter eXpress AVPR1B Activated GPCR Internalization Assay, 10pl Kit. (93-0722E3CP5L)
Catalog No. 502001189
Encompass
The PathHunter AVPR1B Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of AVPR1B via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged AVPR1B. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting AVPR1B-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1189 Supplier Eurofins Discoverx Supplier No. 930722E3CP5L
Add to Cart
Add to Cart
Request a Quote
The PathHunter AVPR1B Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of AVPR1B via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged AVPR1B. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting AVPR1B-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.