Learn More
Eurofins Discoverx InCELL Hunter eXpress Bcl2L1-Bax Protein Binding Assay, 2-plate Kit. (96-0009E3CP16M)
Catalog No. 502004192
Encompass
The InCELL Hunter eXpress Bcl2L1-Bax Protein Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2L1-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2L1 target in an untagged form and Bax binding partner fused to ProLabel. When a compound engages the Bcl2L1 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-4192 Supplier Eurofins Discoverx Supplier No. 960009E3CP16M
The InCELL Hunter eXpress Bcl2L1-Bax Protein Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2L1-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2L1 target in an untagged form and Bax binding partner fused to ProLabel. When a compound engages the Bcl2L1 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.