Learn More
Eurofins Discoverx PathHunter eXpress BMPR1B/ACVR2A Dimerization Assay, 10pl Kit. (93-1010E3CP0L)
Catalog No. 502001833
Encompass
The PathHunter BMPR1B/ACVR2A Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. BMP-6) induced dimerization of BMPR1B/ACVR2A receptors. This product uses EFC technology. The cell line included overexpresses BMPR1B target protein fused to PK alongside ACVR2A target protein fused to EA. Upon ligand-induced dimerization of BMPR1B/ACVR2A, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1833 Supplier Eurofins Discoverx Supplier No. 931010E3CP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter BMPR1B/ACVR2A Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. BMP-6) induced dimerization of BMPR1B/ACVR2A receptors. This product uses EFC technology. The cell line included overexpresses BMPR1B target protein fused to PK alongside ACVR2A target protein fused to EA. Upon ligand-induced dimerization of BMPR1B/ACVR2A, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.