Learn More
Eurofins Discoverx InCELL Hunter eXpress BTK Kinase Binding Assay, 2pl Kit. (96-0056E15CP0M)
Catalog No. 502004436
Encompass
The InCELL Hunter eXpress BTK Kinase Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with kinases. This kit assesses compound-engagement-based stabilization of BTK via detection of BTK levels. This product uses EFC technology. The cell line included overexpresses BTK target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-4436 Supplier Eurofins Discoverx Supplier No. 960056E15CP0M
The InCELL Hunter eXpress BTK Kinase Binding Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with kinases. This kit assesses compound-engagement-based stabilization of BTK via detection of BTK levels. This product uses EFC technology. The cell line included overexpresses BTK target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.