Learn More
Eurofins Discoverx PathHunter eXpress C3AR1 U2OS β-Arrestin GPCR Assay, 2-Plate Kit. (93-0814E3CP0M)
Catalog No. 502001341
Encompass
The PathHunter C3AR1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. C3a) based activation of C3AR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged C3AR1 and EA-tagged β-Arrestin-2. Activation of C3AR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1341 Supplier Eurofins Discoverx Supplier No. 930814E3CP0M
The PathHunter C3AR1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. C3a) based activation of C3AR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged C3AR1 and EA-tagged β-Arrestin-2. Activation of C3AR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.