Learn More
Eurofins Discoverx cAMP Hunter eXpress CALCRL-RAMP1 CHO-K1 GPCR Assay, 2pl Kit. (95-0164E2CP2M)
Catalog No. 502003921
Encompass
The cAMP Hunter CALCRL-RAMP1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. β-CGRP) based activation of CALCRL (paired with RAMP1) GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CALCRL, a naturally Gs coupled GPCR, alongside modulator protein RAMP1. Upon activation of CALCRL, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3921 Supplier Eurofins Discoverx Supplier No. 950164E2CP2M
Add to Cart
Add to Cart
The cAMP Hunter CALCRL-RAMP1 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. β-CGRP) based activation of CALCRL (paired with RAMP1) GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CALCRL, a naturally Gs coupled GPCR, alongside modulator protein RAMP1. Upon activation of CALCRL, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.