Learn More
Eurofins Discoverx cAMP Hunter eXpress CALCRL-RAMP3 CHO-K1 GPCR Assay, 10-plate Kit. (95-0169E2CP2L)
Catalog No. 502003934
Encompass
The cAMP Hunter CALCRL-RAMP3 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. adrenomedullin) based activation of CALCRL (paired with RAMP3) GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CALCRL, a naturally Gs coupled GPCR, alongside modulator protein RAMP3. Upon activation of CALCRL, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3934 Supplier Eurofins Discoverx Supplier No. 950169E2CP2L
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter CALCRL-RAMP3 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. adrenomedullin) based activation of CALCRL (paired with RAMP3) GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CALCRL, a naturally Gs coupled GPCR, alongside modulator protein RAMP3. Upon activation of CALCRL, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.