Learn More
Eurofins Discoverx PathHunter eXpress β-Catenin Nuclear Translocation Assay, 2pl Kit. (93-0743E3CP22M)
Catalog No. 502001229
Encompass
The PathHunter eXpress β-Catenin Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the Wnt-FzGSK3β pathway. This kit assesses ligand (e.g. wnt 3a) based pathway activation via detection of resulting β-Catenin nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged β-Catenin and nuclear localized EA. Upon pathway activation, induced nuclear translocation of β-Catenin leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1229 Supplier Eurofins Discoverx Supplier No. 930743E3CP22M
The PathHunter eXpress β-Catenin Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the Wnt-FzGSK3β pathway. This kit assesses ligand (e.g. wnt 3a) based pathway activation via detection of resulting β-Catenin nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged β-Catenin and nuclear localized EA. Upon pathway activation, induced nuclear translocation of β-Catenin leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.