Learn More
Eurofins Discoverx PathHunter eXpress CDC25A Degradation Assay, 2-Plate Kit. (93-0765E3CP0M)
Catalog No. 502001278
Encompass
The PathHunter eXpress CDC25A Degradation Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting the cell cycle checkpoint pathway. This kit enables detection of levels of CDC25a, allowing for assessment of CHK-1-activation-induced CDC25a degradation. This product uses EFC technology. The cell line included overexpresses CDC25a target protein fused to ProLabel. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of CDC25a EFC signal is lost. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1278 Supplier Eurofins Discoverx Supplier No. 930765E3CP0M
The PathHunter eXpress CDC25A Degradation Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting the cell cycle checkpoint pathway. This kit enables detection of levels of CDC25a, allowing for assessment of CHK-1-activation-induced CDC25a degradation. This product uses EFC technology. The cell line included overexpresses CDC25a target protein fused to ProLabel. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of CDC25a EFC signal is lost. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.