Learn More
Eurofins Discoverx cAMP Hunter eXpress CNR2 CHO-K1 GPCR Assay, 2-Plate Kit. (95-0183E2CP2M)
Catalog No. 502004012
Encompass
The cAMP Hunter CNR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CP 55,940) based activation of CNR2 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CNR2, a naturally Gi/Go coupled GPCR. Upon activation of CNR2, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-4012 Supplier Eurofins Discoverx Supplier No. 950183E2CP2M
The cAMP Hunter CNR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CP 55,940) based activation of CNR2 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type CNR2, a naturally Gi/Go coupled GPCR. Upon activation of CNR2, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.