Learn More
Eurofins Discoverx PathHunter eXpress CSF2RB/CSF2RA Dimerization Assay, 2pl Kit. (93-1078E3CP22M)
Catalog No. 502001991
Encompass
The PathHunter CSF2RB/CSF2RA Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. GM-CSF) induced dimerization of CSF2RB/CSF2RA receptors. This product uses EFC technology. The cell line included overexpresses CSF2RB target protein fused to PK alongside CSF2RA target protein fused to EA. Upon ligand-induced dimerization of CSF2RB/CSF2RA, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1991 Supplier Eurofins Discoverx Supplier No. 931078E3CP22M
The PathHunter CSF2RB/CSF2RA Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. GM-CSF) induced dimerization of CSF2RB/CSF2RA receptors. This product uses EFC technology. The cell line included overexpresses CSF2RB target protein fused to PK alongside CSF2RA target protein fused to EA. Upon ligand-induced dimerization of CSF2RB/CSF2RA, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.