Learn More
Eurofins Discoverx PathHunter eXpress CXCR6 HEK 293 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0205E1CP0L)
Catalog No. 501999943
Encompass
The PathHunter CXCR6 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CXCL16) based activation of CXCR6 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR6 and EA-tagged β-Arrestin-2. Activation of CXCR6 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-199-9943 Supplier Eurofins Discoverx Supplier No. 930205E1CP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter CXCR6 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CXCL16) based activation of CXCR6 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CXCR6 and EA-tagged β-Arrestin-2. Activation of CXCR6 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.