Learn More
Eurofins Discoverx PathHunter eXpress cyno CXCR2 HEK 293 β-Arrestin GPCR Assay, 10pl Kit. (93-0981E1CCP0L)
Catalog No. 502001738
Encompass
The PathHunter cyno CXCR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CXCL8/IL8) based activation of cyno CXCR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno CXCR2 and EA-tagged β-Arrestin-2. Activation of cyno CXCR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Cynomolgus monkey version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1738 Supplier Eurofins Discoverx Supplier No. 930981E1CCP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter cyno CXCR2 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. CXCL8/IL8) based activation of cyno CXCR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno CXCR2 and EA-tagged β-Arrestin-2. Activation of cyno CXCR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Cynomolgus monkey version of this protein. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.