Learn More
Eurofins Discoverx PathHunter eXpress cyno GPR120 CHO-K1 β-Arrestin GPCR Assay, 10pl Kit. (93-1029E2CP0L)
Catalog No. 502001881
Encompass
The PathHunter cyno GPR120 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. free fatty acids) based activation of cyno GPR120 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno GPR120 and EA-tagged β-Arrestin-2. Activation of cyno GPR120 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1881 Supplier Eurofins Discoverx Supplier No. 931029E2CP0L
Add to Cart
Add to Cart
Request a Quote
The PathHunter cyno GPR120 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. free fatty acids) based activation of cyno GPR120 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno GPR120 and EA-tagged β-Arrestin-2. Activation of cyno GPR120 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.