Learn More
Optimize Technologies EXP™ Direct Connect Holder Kit w/ EXP™2 TI-LOK Fitting
SDP
Catalog No. 502006353
Encompass_Preferred
The EXP™ Direct Connect Holder is rated to 20,000psi with hand-tight replaceable cartridges. This unique design connects directly to any UHPLC column with the auto-adjusting Titanium Hybrid Ferrule and EXP™ Fittings, providing repeated zero-dead-volume connections and easy adjustability between various column types. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/ outlet. The combination of low-dispersion and ultra high-pressure capabilities provides effective protection with negligible effects on retention time, plate count, and peak shape. The holder features the Optimize TI-LOK fitting: a one piece nut and ferrule design that prevents the user from wasting time and money looking for lost ferrules. Use with EXP™ Trap and Guard Cartridges.
Catalog No. 50-200-6353 Supplier Optimize Technologies Supplier No. 150205243
The EXP™ Direct Connect Holder is rated to 20,000psi with hand-tight replaceable cartridges. This unique design connects directly to any UHPLC column with the auto-adjusting Titanium Hybrid Ferrule and EXP™ Fittings, providing repeated zero-dead-volume connections and easy adjustability between various column types. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/ outlet. The combination of low-dispersion and ultra high-pressure capabilities provides effective protection with negligible effects on retention time, plate count, and peak shape. The holder features the Optimize TI-LOK fitting: a one piece nut and ferrule design that prevents the user from wasting time and money looking for lost ferrules. Use with EXP™ Trap and Guard Cartridges.