Learn More
Eurofins Discoverx PathHunter eXpress EDG4 CHO-K1 β-Arrestin GPCR Assay, 10-Plate Kit. (93-0255E2CP13L)
Catalog No. 502000093
Encompass
The PathHunter EDG4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. LPA) based activation of EDG4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG4 and EA-tagged β-Arrestin-2. Activation of EDG4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0093 Supplier Eurofins Discoverx Supplier No. 930255E2CP13L
Add to Cart
Add to Cart
Request a Quote
The PathHunter EDG4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. LPA) based activation of EDG4 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG4 and EA-tagged β-Arrestin-2. Activation of EDG4 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.