Learn More
Eurofins Discoverx PathHunter eXpress EGFR/EGFR Dimerization Assay, 10-Plate Kit. (93-0989E3CP5L)
Catalog No. 502001760
Encompass
The PathHunter EGFR/EGFR Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. EGF) induced dimerization of EGFR/EGFR receptors. This product uses EFC technology. The cell line included overexpresses EGFR target protein fused to PK alongside EGFR target protein fused to EA. Upon ligand-induced dimerization of EGFR/EGFR, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1760 Supplier Eurofins Discoverx Supplier No. 930989E3CP5L
Add to Cart
Add to Cart
Request a Quote
The PathHunter EGFR/EGFR Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. EGF) induced dimerization of EGFR/EGFR receptors. This product uses EFC technology. The cell line included overexpresses EGFR target protein fused to PK alongside EGFR target protein fused to EA. Upon ligand-induced dimerization of EGFR/EGFR, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.