Learn More
Eurofins Discoverx PathHunter eXpress EpoR/EpoR Dimerization Assay, 2-Plate Kit. (93-0965E3CP0M)
Catalog No. 502001694
Encompass
The PathHunter EpoR/EpoR Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. erythropoietin) induced dimerization of EpoR/EpoR receptors. This product uses EFC technology. The cell line included overexpresses EpoR target protein fused to PK alongside EpoR target protein fused to EA. Upon ligand-induced dimerization of EpoR/EpoR, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1694 Supplier Eurofins Discoverx Supplier No. 930965E3CP0M
The PathHunter EpoR/EpoR Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. erythropoietin) induced dimerization of EpoR/EpoR receptors. This product uses EFC technology. The cell line included overexpresses EpoR target protein fused to PK alongside EpoR target protein fused to EA. Upon ligand-induced dimerization of EpoR/EpoR, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.