Learn More
Eurofins Discoverx PathHunter eXpress EpoR-JAK2 Functional Assay, 10-Plate Kit. (93-0900E3CP16L)
Catalog No. 502001524
Encompass
The PathHunter EpoR-JAK2 Functional assay eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting CTKs. This kit assesses EpoR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged EpoR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1524 Supplier Eurofins Discoverx Supplier No. 930900E3CP16L
Add to Cart
Add to Cart
Request a Quote
The PathHunter EpoR-JAK2 Functional assay eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting CTKs. This kit assesses EpoR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged EpoR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.