Learn More
Eurofins Discoverx PathHunter eXpress ErbB2/ErbB4 Dimerization Assay, 2pl Kit. (93-0960E3CP5M)
Catalog No. 502001675
Encompass
The PathHunter ErbB2/ErbB4 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. heregulin β-1) induced dimerization of ErbB2/ErbB4 receptors. This product uses EFC technology. The cell line included overexpresses ErbB2 target protein fused to PK alongside ErbB4 target protein fused to EA. Upon ligand-induced dimerization of ErbB2/ErbB4, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1675 Supplier Eurofins Discoverx Supplier No. 930960E3CP5M
The PathHunter ErbB2/ErbB4 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. heregulin β-1) induced dimerization of ErbB2/ErbB4 receptors. This product uses EFC technology. The cell line included overexpresses ErbB2 target protein fused to PK alongside ErbB4 target protein fused to EA. Upon ligand-induced dimerization of ErbB2/ErbB4, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.