Learn More
Eurofins Discoverx PathHunter eXpress F2R (PAR1) CHO-K1 β-Arrestin GPCR Assay, 2pl Kit. (93-0278E2CP0M)
Catalog No. 502000156
Encompass
The PathHunter F2R (PAR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. proteases, TFLLRN) based activation of F2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged F2R and EA-tagged β-Arrestin-2. Activation of F2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0156 Supplier Eurofins Discoverx Supplier No. 930278E2CP0M
The PathHunter F2R (PAR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. proteases, TFLLRN) based activation of F2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged F2R and EA-tagged β-Arrestin-2. Activation of F2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.