Learn More
Eurofins Discoverx PathHunter eXpress FGFR4-β-Klotho Functional Assay, 2pl Kit. (93-1060E3CP0M)
Catalog No. 502001957
Encompass
The PathHunter eXpress FGFR4-β-Klotho Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated FGFR4 and β-Klotho dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged FGFR4 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1957 Supplier Eurofins Discoverx Supplier No. 931060E3CP0M
The PathHunter eXpress FGFR4-β-Klotho Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated FGFR4 and β-Klotho dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged FGFR4 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.