Learn More
Eurofins Discoverx PathHunter eXpress FOXO3 Nuclear Translocation Assay, 10pl Kit. (93-0876E3CP5L)
Catalog No. 502001473
Encompass
The PathHunter eXpress FOXO3 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the PI3 pathway. This kit assesses ligand (e.g. wortmannin) based pathway activation via detection of resulting FOXO3 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged FOXO3 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of FOXO3 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1473 Supplier Eurofins Discoverx Supplier No. 930876E3CP5L
Add to Cart
Add to Cart
Request a Quote
The PathHunter eXpress FOXO3 Nuclear Translocation Assay provides an easy-to-use cell based assay to study drug candidates affecting the PI3 pathway. This kit assesses ligand (e.g. wortmannin) based pathway activation via detection of resulting FOXO3 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged FOXO3 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of FOXO3 leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.