Learn More
Eurofins Discoverx PathHunter eXpress GALR1 Total Internalization Assay, 2pl Kit. (93-0781E3CP5M)
Catalog No. 502001300
Encompass
The PathHunter GALR1 Total Internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of GALR1 via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged GALR1 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GALR1 receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1300 Supplier Eurofins Discoverx Supplier No. 930781E3CP5M
Add to Cart
Add to Cart
The PathHunter GALR1 Total Internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of GALR1 via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged GALR1 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GALR1 receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.