Learn More
Eurofins Discoverx PathHunter eXpress GHR-JAK2 Functional Assay, 2-Plate Kit. (93-0829E3CP14M)
Catalog No. 502001379
Encompass
The PathHunter GHR-JAK2 Functional assay eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting CTKs. This kit assesses GHR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged GHR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1379 Supplier Eurofins Discoverx Supplier No. 930829E3CP14M
The PathHunter GHR-JAK2 Functional assay eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting CTKs. This kit assesses GHR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged GHR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.