Learn More
Eurofins Discoverx cAMP Hunter eXpress GHRHR CHO-K1 GPCR Assay, 2-Plate Kit. (95-0076E2CP2M)
Catalog No. 502002726
Encompass
The cAMP Hunter GHRHR eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. GRF) based activation of GHRHR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GHRHR, a naturally Gs coupled GPCR. Upon activation of GHRHR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2726 Supplier Eurofins Discoverx Supplier No. 950076E2CP2M
The cAMP Hunter GHRHR eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. GRF) based activation of GHRHR GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GHRHR, a naturally Gs coupled GPCR. Upon activation of GHRHR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.