Learn More
Eurofins Discoverx PathHunter eXpress GHSR1a U2OS β-Arrestin-1 GPCR Assay, 2-Plate Kit. (93-0242E3CP5M)
Catalog No. 502000057
Encompass
The PathHunter GHSR1a (ARR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. ghrelin) based activation of GHSR1a GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1a and EA-tagged β-Arrestin-1. Activation of GHSR1a stimulates the recruitment of β-Arrestin-1 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0057 Supplier Eurofins Discoverx Supplier No. 930242E3CP5M
The PathHunter GHSR1a (ARR1) eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. ghrelin) based activation of GHSR1a GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1a and EA-tagged β-Arrestin-1. Activation of GHSR1a stimulates the recruitment of β-Arrestin-1 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.