Learn More
Eurofins Discoverx PathHunter eXpress GHSR1b CHO-K1 β-Arrestin Orphan GPCR Assay, 2-Plate Kit. (93-0354E2ACP1M)
Catalog No. 502000334
Encompass
The PathHunter GHSR1b eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. ghrelin) based activation of GHSR1b GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1b and EA-tagged β-Arrestin-2. Activation of GHSR1b stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0334 Supplier Eurofins Discoverx Supplier No. 930354E2ACP1M
Add to Cart
Add to Cart
The PathHunter GHSR1b eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. ghrelin) based activation of GHSR1b GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR1b and EA-tagged β-Arrestin-2. Activation of GHSR1b stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.