Learn More
Eurofins Discoverx cAMP Hunter eXpress GLP1R CHO-K1 GPCR Assay, 10-Plate Kit. (95-0062E2CP2L)
Catalog No. 502002601
Encompass
The cAMP Hunter GLP1R eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. exendin-4) based activation of GLP1R GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GLP1R, a naturally Gs coupled GPCR. Upon activation of GLP1R, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2601 Supplier Eurofins Discoverx Supplier No. 950062E2CP2L
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter GLP1R eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. exendin-4) based activation of GLP1R GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GLP1R, a naturally Gs coupled GPCR. Upon activation of GLP1R, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.