Learn More
Eurofins Discoverx cAMP Hunter eXpress GPR109A CHO-K1 GPCR Assay, 2-Plate Kit. (95-0122E2CP2M)
Catalog No. 502003322
Encompass
The cAMP Hunter GPR109A eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. nicotinic acid) based activation of GPR109A GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPR109A, a naturally Gi/Go coupled GPCR. Upon activation of GPR109A, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3322 Supplier Eurofins Discoverx Supplier No. 950122E2CP2M
The cAMP Hunter GPR109A eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. nicotinic acid) based activation of GPR109A GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPR109A, a naturally Gi/Go coupled GPCR. Upon activation of GPR109A, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.