Learn More
Eurofins Discoverx cAMP Hunter eXpress GPR109B CHO-K1 GPCR Assay, 10-Plate Kit. (95-0141E2CP2L)
Catalog No. 502003658
Encompass
The cAMP Hunter GPR109B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. 3-hydroxyoctanoic acid) based activation of GPR109B GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPR109B, a naturally Gi/Go coupled GPCR. Upon activation of GPR109B, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-3658 Supplier Eurofins Discoverx Supplier No. 950141E2CP2L
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter GPR109B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. 3-hydroxyoctanoic acid) based activation of GPR109B GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GPR109B, a naturally Gi/Go coupled GPCR. Upon activation of GPR109B, the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.