Learn More
Eurofins Discoverx PathHunter eXpress GPR120S (Short Isoform) CHO-K1 β-Arrestin GPCR Assay, 2-plate Kit. (93-0924E2CP0M)
Catalog No. 502001570
Encompass
The PathHunter GPR120S eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. free fatty acids) based activation of GPR120S GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR120S and EA-tagged β-Arrestin-2. Activation of GPR120S stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1570 Supplier Eurofins Discoverx Supplier No. 930924E2CP0M
The PathHunter GPR120S eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. free fatty acids) based activation of GPR120S GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR120S and EA-tagged β-Arrestin-2. Activation of GPR120S stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.