Learn More
Eurofins Discoverx PathHunter eXpress GPR15 CHO-K1 β-Arrestin Orphan GPCR Assay, 2-Plate Kit. (93-0682E2CP1M)
Catalog No. 502001096
Encompass
The PathHunter GPR15 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR15 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR15 and EA-tagged β-Arrestin-2. Activation of GPR15 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. This is an orphan receptor without a known ligand. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1096 Supplier Eurofins Discoverx Supplier No. 930682E2CP1M
The PathHunter GPR15 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR15 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR15 and EA-tagged β-Arrestin-2. Activation of GPR15 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. This is an orphan receptor without a known ligand. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.