Learn More
Eurofins Discoverx PathHunter eXpress GPR79 CHO-K1 β-Arrestin Orphan GPCR Assay, 10-Plate Kit. (93-0408E2ACP1L)
Catalog No. 502000470
Encompass
The PathHunter GPR79 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR79 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR79 and EA-tagged β-Arrestin-2. Activation of GPR79 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. This is an orphan receptor without a known ligand. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-0470 Supplier Eurofins Discoverx Supplier No. 930408E2ACP1L
Add to Cart
Add to Cart
Request a Quote
The PathHunter GPR79 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR79 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR79 and EA-tagged β-Arrestin-2. Activation of GPR79 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. This is an orphan receptor without a known ligand. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.