Learn More
Eurofins Discoverx InCELL Hunter eXpress HEK 293 BRD3(1,2) Bromodomain Assay, 2-plate Kit. (96-0038E1CP7M)
Catalog No. 502004244
Encompass
The InCELL Hunter eXpress HEK 293 BRD3(1,2) Bromodomain Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with regulators of epigenetics pathways. This kit assesses compound-engagement-based stabilization of BRD3(1,2) via detection of BRD3(1,2) levels. This product uses EFC technology. The cell line included overexpresses BRD3(1,2) target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-4244 Supplier Eurofins Discoverx Supplier No. 960038E1CP7M
The InCELL Hunter eXpress HEK 293 BRD3(1,2) Bromodomain Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with regulators of epigenetics pathways. This kit assesses compound-engagement-based stabilization of BRD3(1,2) via detection of BRD3(1,2) levels. This product uses EFC technology. The cell line included overexpresses BRD3(1,2) target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.