Learn More
Eurofins Discoverx InCELL Hunter eXpress HEK 293 BRDT(1,2) Bromodomain Assay, 10-plate Kit. (96-0046E1CP7L)
Catalog No. 502004318
Encompass
The InCELL Hunter eXpress HEK 293 BRDT(1,2) Bromodomain Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with regulators of epigenetics pathways. This kit assesses compound-engagement-based stabilization of BRDT(1,2) via detection of BRDT(1,2) levels. This product uses EFC technology. The cell line included overexpresses BRDT(1,2) target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-4318 Supplier Eurofins Discoverx Supplier No. 960046E1CP7L
The InCELL Hunter eXpress HEK 293 BRDT(1,2) Bromodomain Assay provides an easy-to-use cell based assay to study small molecule drug candidates that engage with regulators of epigenetics pathways. This kit assesses compound-engagement-based stabilization of BRDT(1,2) via detection of BRDT(1,2) levels. This product uses EFC technology. The cell line included overexpresses BRDT(1,2) target protein fused to ProLabel. In the absence of compound that binds and stabilizes the target, this protein is degraded and no EFC signal is produced. When compound engages and stabilizes the target, EA contained in the detection reagent produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.