Learn More
Eurofins Discoverx PathHunter eXpress HTR1B U2OS β-Arrestin GPCR Assay, 10-Plate Kit. (93-0697E3CP6L)
Catalog No. 502001140
Encompass
The PathHunter HTR1B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. serotonin) based activation of HTR1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1B and EA-tagged β-Arrestin-2. Activation of HTR1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1140 Supplier Eurofins Discoverx Supplier No. 930697E3CP6L
Add to Cart
Add to Cart
Request a Quote
The PathHunter HTR1B eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. serotonin) based activation of HTR1B GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged HTR1B and EA-tagged β-Arrestin-2. Activation of HTR1B stimulates the recruitment of β-Arrestin-2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.