Learn More
Eurofins Discoverx PathHunter eXpress HTR2C Activated GPCR Internalization Assay, 2pl Kit. (93-0669E3CP27M)
Catalog No. 502001069
Encompass
The PathHunter HTR2C Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of HTR2C via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged HTR2C. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting HTR2C-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-1069 Supplier Eurofins Discoverx Supplier No. 930669E3CP27M
The PathHunter HTR2C Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of HTR2C via detection of localization to the early endosome. The included cell line overexpresses EA-tagged β-Arrestin-2 as well as a ProLink-tagged early endosome protein and untagged HTR2C. Upon ligand-induced receptor activation, β-Arrestin-2 is recruited to the activated GPCR and the resulting HTR2C-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.