Learn More
Eurofins Discoverx PathHunter eXpress IFNGR1/IFNGR2 Dimerization Assay, 2pl Kit. (93-1098E3CP5M)
Catalog No. 502002073
Encompass
The PathHunter IFNGR1/IFNGR2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IFN-g) induced dimerization of IFNGR1/IFNGR2 receptors. This product uses EFC technology. The cell line included overexpresses IFNGR1 target protein fused to PK alongside IFNGR2 target protein fused to EA. Upon ligand-induced dimerization of IFNGR1/IFNGR2, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. 50-200-2073 Supplier Eurofins Discoverx Supplier No. 931098E3CP5M
The PathHunter IFNGR1/IFNGR2 Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IFN-g) induced dimerization of IFNGR1/IFNGR2 receptors. This product uses EFC technology. The cell line included overexpresses IFNGR1 target protein fused to PK alongside IFNGR2 target protein fused to EA. Upon ligand-induced dimerization of IFNGR1/IFNGR2, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.